Tlačiť
Weighing bottles, glass
Dodávateľ: WINZER LABORGLASTECHN
20-116-50 20-112-40 20-112-44 20-112-46 20-112-50 20-112-60
WINZ20-116-50EA 0 EUR
WINZ20-116-50 WINZ20-112-40 WINZ20-112-44 WINZ20-112-46 WINZ20-112-50 WINZ20-112-60
Weighing bottles, glass
Láhve Váženky
Borosilicate glass 3,3.
  • With interchangeable cap
  • Exterior joint improves material transfers and reduces risk of material contamination of cap
  • Available in low or tall forms
With interchangeable standard ground cap and blank label with size indication.