Tlačiť
Safety shoes, lace-up, Concept Plus, Provoke
Dodávateľ: U GROUP
UC20064/38 UC20064/39 UC20064/40 UC20064/41 UC20064/42 UC20064/43 UC20064/44 UC20064/45 UC20064/46 UC20064/47 UC20064/48
UPOWUC20064/38EA 121.54 EUR
UPOWUC20064/38 UPOWUC20064/39 UPOWUC20064/40 UPOWUC20064/41 UPOWUC20064/42 UPOWUC20064/43 UPOWUC20064/44 UPOWUC20064/45 UPOWUC20064/46 UPOWUC20064/47 UPOWUC20064/48
Safety shoes, lace-up, Concept Plus, Provoke
Obuv
Metal-free safety shoes made from water repellent full grain leather and a hard wearing, anti-static, antislip PU sole.
  • Light non-slip Actiondry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Airtoe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Brown
Certifikácie: EN 20345:2011; S3, SRC