Tlačiť
Temperature indicating kit
Dodávateľ: KLEINFELD
1216165 1211103 1216168
KLEI1216165EA 503.01 EUR
KLEI1216165 KLEI1211103 KLEI1216168
Temperature indicating kit
Proužky indikátorů teploty
These Thermax chill checker include eight different measurement points over a temperature range from −17 to +20 °C. Activation takes place after cooling by pressing in the center. When the temperature is exceeded, it changes color purple or blue.