Tlačiť
Spill control pallets, EcoPolyBlend™
Dodávateľ: JUSTRITE MANUFACTURING
28635 28623 28631
119-0378EA 616.53 EUR
119-0378 119-0376 119-0377
Spill control pallets, EcoPolyBlend™
Skladování chemikálií
These pallets are made of up to 100% recycled polyethylene; dirt hiding black colour is 100% recycled, high visibility yellow is 35% recycled content. Fully compliant and quality engineered with a robust design, they withstand the toughest environments and are nearly impervious to chemical attack.
  • Independently validated by UL Environment
  • Seamless, robust design eliminates leaks
  • Excellent chemical compatibility and no corrosion
  • Sturdy and self-locating grates remove for easy cleaning
  • Square or space saving linear styles in multiple sizes offer storage versatility
Certifikácie: Meets or exceeds EPA, SPCC, NFPA 1, and the Int’l Fire Code.