Tlačiť
Precision hydrometers, ASTM (API Grade)
Dodávateľ: SCHNEIDER LUDWIG
3202010 3202009 3202008 3202007 3202006 3202005 3202004 3202003 3202002 3202001
SCNL3202010EA 60.97 EUR
SCNL3202010 SCNL3202009 SCNL3202008 SCNL3202007 SCNL3202006 SCNL3202005 SCNL3202004 SCNL3202003 SCNL3202002 SCNL3202001
Precision hydrometers, ASTM (API Grade)
Hydrometry
These precision hydrometers are calibrated without thermometer.
Error limit: ±0,1° API
Reference temperature: 15,5 °C
Certifikácie: American Petroleum Institute (API) and ASTM E100-10.
Balenie: Each packed in a square plastic sleeve.