Tlačiť
Safety wash bottles, narrow neck, with printed name
Dodávateľ: MRAZ EDWIN
2506/500 2506/1000 2500/500 2500/1000 2501/500 2501/1000 2505/250 2505/500 2505/1000 2504/1000 2503/250 2503/500 2503/1000 2502/500 2502/1000 2509/500
215-6902EA 5.78 EUR
215-6902 215-6903 215-6905 215-6906 215-6908 215-6909 215-6914 215-6915 215-6916 215-6919 215-6921 215-6922 215-6923 215-6925 215-6926 215-6951
Safety wash bottles, narrow neck, with printed name
Láhve Střičky
PE, yellow coloured.
  • Yellow coloured for largely light absorbing
  • Good chemical resistance, good temperature resistance
With imprint of capacityand hazard identification according to the regualtions for the handling of dangerous materials.