Tlačiť
Váženka na žíraviny
Dodávateľ: TECHNOSKLO
632421203051 632421203052
TNOS632421203051EA 49.67 EUR
TNOS632421203051 TNOS632421203052
Váženka na žíraviny
Láhve Váženky
Made of borosilicate glass.
  • Excellent chemical resistance
  • Excellent temperature resistance
With stopcock with glass key or valve with PTFE spindle.