Tlačiť
NMR tubes, high-throughput, thin walled, 5 mm Ø, SP Wilmad-LabGlass
Dodávateľ: SP Industries
WG-1000-7 WG-1000-8 WG-1000-4
634-0871EA 225.03 EUR
634-0871 634-0872 634-1157
NMR tubes, high-throughput, thin walled, 5 mm Ø, SP Wilmad-LabGlass
Zkumavky Zkumavky pro NMR
Borosilicate glass, class B, ASTM E438 Type 1
  • Best Ø tolerance in the industry and a 30% thicker wall than any competitors' product
  • Manufactured in an ISO 9001:2008 USA facility
  • Camber and concentricity values listed are the Total Indicator Reading (TIR)
  • 100% visual and physical inspection ensures quality, including physical dimensions and surface defects
  • With or without disposable cap
Thin walled high throughput NMR tubes have an average camber of 60 µm to guarantee the clarity of the spectrum for small molecule (MW<250) samples up to 600 MHz.
Pozor: Recommended only for experiments with small organic molecules (Molecular Weight ~500) at ambient temperatures only. Cooling/heating of these tubes may lead to breakage; for NMR experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, refer to Wilmad Precision NMR Tubes.