Tlačiť
Impact testers
Dodávateľ: Industrial Physics
SP1891 SP1890
SIMESP1891EA 3082.23 EUR
SIMESP1891 SIMESP1890
Impact testers
Testery nátěrů
Impact testers used to determine the impact resistivity and flexibility of materials.
  • Spirit level built-in for correct positioning
  • Dual scale instrument equipped with a special guidance
  • Distance between each impact is always according to the standards
Certifikácie: DIN 6272-1 and -2, ASTM D2794 and G14.