Tlačiť
VWR®, Test Sieves, 300×60 mm
Dodávateľ: VWR Collection
510-2697EA 294.42 EUR
510-2697 510-2698 510-2699 510-2700 510-2701 510-2702 510-2703 510-2704 510-2705 510-2706 510-2707 510-2708 510-2709 510-2710 510-2711 510-2712 510-2713 510-2714 510-2715 510-2716 510-2717 510-2718 510-2719 510-2720 510-2721 510-2722 510-2723 510-2724 510-2725 510-2726 510-2727 510-2728 510-2729 510-2730 510-2731 510-2732 510-2733 510-2734 510-2735 510-2736 510-2737 510-2738 510-2739 510-2740 510-2741 510-2742 510-2743 510-2744 510-2745 510-2746 510-2747 510-2748 510-2749 510-2750 510-2751 510-2752 510-2753 510-2754 510-2755 510-2756 510-2757 510-2758 510-2759 510-2760 510-2761 510-2762 510-2763 510-2764 510-2765 510-2766 510-2767 510-2768 510-2769 510-2770 510-2771 510-2772 510-2773 510-2774 510-2775 510-2776 510-2777 510-2778 510-2779 510-2780 510-2781 510-2782 510-2783 510-2784 510-2785 510-2786 510-2787 510-2788
VWR®, Test Sieves, 300×60 mm
Síta Síta
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Informace o dodávce: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.