Tlačiť
VWR®, Test Sieves, 76×22 mm
Dodávateľ: VWR Collection
510-3611EA 992.78 EUR
510-3611 510-3612 510-3613 510-3614 510-3615 510-3616 510-3617 510-3618 510-3619 510-3620 510-3621 510-3622 510-3623 510-3624 510-3625 510-3626 510-3627 510-3628 510-3629 510-3630 510-3631 510-3632 510-3633 510-3634 510-3635 510-3636 510-3637 510-3638 510-3639 510-3640 510-3641 510-3642 510-3643 510-3644 510-3645 510-3646 510-3647 510-3648 510-3649 510-3650 510-3651 510-3652 510-3653 510-3654 510-3655 510-3656 510-3657 510-3658 510-3659 510-3660 510-3661 510-3814 510-3815 510-3816
VWR®, Test Sieves, 76×22 mm
Síta Síta
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Informace o dodávce: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.