Tlačiť
VWR®, Test Sieves, 315×75 mm
Dodávateľ: VWR Collection
510-1094EA 307.66 EUR
510-1094 510-1095 510-1096 510-1097 510-1098 510-1099 510-1101 510-1102 510-1103 510-1104 510-1105 510-1106 510-1107 510-1108 510-1109 510-1180 510-1181 510-1182 510-1183 510-1184 510-1185 510-1186 510-1110 510-1111 510-1112 510-1113 510-1114 510-1115 510-1116 510-1117 510-1118 510-1119 510-1120 510-1121 510-1122 510-1123 510-1124 510-1125 510-1126 510-1127 510-1128 510-1129 510-1130 510-1131 510-1132 510-1133 510-1134 510-1135 510-1136 510-1137 510-1138 510-1139 510-1140 510-1141 510-1142 510-1143 510-1144 510-1145 510-1146 510-1147 510-1148 510-1149 510-1150 510-1151 510-1152 510-1153 510-1154 510-1155 510-1156 510-1157 510-1158 510-1159 510-1160 510-1161 510-1162 510-1163 510-1164 510-1165 510-1166 510-1167 510-1168 510-1169 510-1170 510-1171 510-1172 510-1173 510-1174 510-1175 510-1176 510-1177 510-1178 510-1179
VWR®, Test Sieves, 315×75 mm
Síta Síta
Frame: Stainless steel; woven wire cloth: Stainless steel, ISO 3310, ASTM E11, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Informace o dodávce: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.