Tlačiť
Safety underbench cabinets, type 90, UTS ergo line S, 600 mm depth
Dodávateľ: DUPERTHAL
29-060669-503 29-060669-403 29-060669-003 29-060667-403 29-060667-003 29-060667-503
DUEH29-060669-503EA 1760.87 EUR
DUEH29-060669-503 DUEH29-060669-403 DUEH29-060669-003 135-1896 DUEH29-060667-003 DUEH29-060667-503
Safety underbench cabinets, type 90, UTS ergo line S, 600 mm depth
Nábytek kancelářský Skříně pro zásoby
These safety cabinets are fire resistant up to 90 minutes. These under bench cabinets are equipped with wing doors and/or drawers for the storage of hazardous substances in working spaces.
  • Easy moving, single-handed operation of the doors and drawers
  • All UTS ergo line models are fitted with a cylinder lock as a standard
  • Two door technologies are available: the classical on or the push-to open wing door technology (the easy moving safety doors are opened by applying a gentle push to the front)
Drawers or wing doors remain open in any position and containers can be inserted and removed using both hands. The drawers/doors close automatically in case of fire. All UTS models come with standard right hand door hinge, optionally available with left hand door hinge (on request).
Standard equipotential bonding saddle on the roof of the cabinet for connecting to the earthing in accordance with TRBS 2153, BGR 132 (avoiding ignition hazards). The interior fittings are conductively connected with the equipotential bonding saddle.
Constant floor extraction by the slotted base (height 35 mm). This enables harmful substances or vapours which collect at the bottom to be safely picked up and removed with the exhaust air. The exhaust connection Ø is 75 mm on the rear of the cabinet. The cut-off flaps close automatically in the case of fire.
Wing doors and carcass made from sheet steel, powder-coated. Interior fittings made from powder-coated sheet steel. One pull-out tray, one perforated sheet insert and classical base 35 mm.
In accordance with TRbF 20 Annex L. Type testing according to DIN EN 14470-1 and DIN EN 14727 (laboratory furniture standard) by recognised test institutes (TÜV Süd, ift Rosenheim) and classification type 90 in accordance with DIN EN 14470-1, GS/CE mark.
Certifikácie: In accordance with TRbF 20 Annex L. Type testing according to DIN EN 14470-1 and DIN EN 14727 (laboratory furniture standard) by recognised test institutes (TÜV Süd, ift Rosenheim) and classification type 90 in accordance with DIN EN 14470-1, GS/CE mark.
Informace o dodávce: Delivery is not included, prices available upon request.