Tlačiť
VWR®, Test Sieves, 350×60 mm, with Square Holes
Dodávateľ: VWR Collection
510-3430EA 346.57 EUR
510-3430 510-3431 510-3432 510-3433 510-3434 510-3435 510-3436 510-3437 510-3438 510-3439 510-3440 510-3441 510-3442 510-3443 510-3444 510-3445 510-3446 510-3447 510-3448 510-3449 510-3450 510-3451 510-3452 510-3453 510-3454 510-3455 510-3456 510-3457 510-3458 510-3459 510-3460 510-3461 510-3462 510-3463 510-3464 510-3465 510-3466 510-3467 510-3468 510-3469 510-3470
VWR®, Test Sieves, 350×60 mm, with Square Holes
Síta Síta
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Informace o dodávce: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.