Tlačiť
VWR®, Test Sieves, 200×50 mm, with Square Holes
Dodávateľ: VWR Collection
510-0799EA 204.97 EUR
510-0799 510-0800 510-0801 510-0802 510-0803 510-0804 510-0805 510-0806 510-0807 510-0808 510-0809 510-0810 510-0811 510-0812 510-0813 510-0814 510-0815 510-0816 510-0817 510-0818 510-0819 510-0820 510-0821 510-0822 510-0823 510-0824 510-0825 510-0826 510-0827 510-0828 510-0829 510-0830 510-0831 510-0832 510-0833 510-0834 510-0835 510-0836 510-0837 510-0838 510-0839
VWR®, Test Sieves, 200×50 mm, with Square Holes
Síta Síta
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.
Informace o dodávce: Supplied as standard with a certificate of compliance 2.1 according to DIN EN 10204.