Leptin is secreted by white adipocytes and functions as part of a signaling pathway that can inhibit food intake and/or regulate energy expenditure to maintain constancy of the adipose mass. Leptin also has several endocrine functions and is involved in the regulation of immune and inflammatory responses, hematopoiesis, angiogenesis and wound healing (Ref: Entrez Gene).
The antibody reacts with human leptin protein and may react with other speces leptin because of protein homology but is currently untested.
Application Information:
ELISA and WB. Suggested dilution of 1:1,000-1:2,000. Biosensis recommends that the optimal working dilution should be determined by the end user.
Type: Primary
Antigen: Leptin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Multiple points of reactivity likely because of the polyclonal nature of the antibody
Host: Chicken
Isotype: IgY
Reactivity: Human