Tlačiť
Safety wash bottles, wide neck, multilingual, Azlon®
Dodávateľ: DWK LIFE SCIENCES
WGW701PP WGW703PP
215-3791EA 56.56 EUR
215-3791 215-3792
Safety wash bottles, wide neck, multilingual, Azlon®
Láhve Střičky
LDPE, translucent, with PP spray cap.
  • Indelibly printed in four languages (English, French, German and Spanish)
  • Solvent labels including the new Globally Harmonised Symbols, NFPA diamond, risk phrases and safety information
  • Closure feature precision jet tip aperture for fine control
Wide neck bottles fitted with colour coded one-piece PP closure that maintain their shape in use.
Certifikácie: Azlon® wash bottles meet the material requirements of the U.S. Food and Drug Administration as specified in 21 CFR 177.1520.