Tlačiť

Anti-Glutathione S-Transferase Mouse Monoclonal Antibody [clone: 5C11 (6C7)]

Dodávateľ: UNITED BIOCHEMICALS
G8135-06-1 G8135-06-500
USBIG8135-06-1EA 1091.53 EUR
USBIG8135-06-1 USBIG8135-06-500
Anti-Glutathione S-Transferase Mouse Monoclonal Antibody [clone: 5C11 (6C7)]
Protilátky
Type: Tag
Antigen: Glutathione S-Transferase
Clonality: Monoclonal
Clone: 5C11 (6C7)
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR