Tlačiť
Autosampler vials, Supelco®
Dodávateľ: Merck
854165 SU860033
SIAL854165EA 26.47 EUR
SIAL854165 SIALSU860033
Autosampler vials, Supelco®
Vialky
These short thread autosampler vials with 9 mm thread are designed to fit almost all autosamplers.