Tlačiť
Grease, vacuum, Apiezon®
Dodávateľ: Edwards
Apiezon®
EDWAH11150001EA 0 EUR
EDWAH11150001
Grease, vacuum, Apiezon®
Lubrikanty
A range of Apiezon® greases is offered for use on high vacuum systems and for many general applications in the laboratory. These high purity greases are non-toxic, are easy to apply and clean off and have long shelf lives.
  • With good glittering qualities
  • Non-vacuum uses include lubrication of glass and metal taps in laboratory, medical and industrial applications