Tlačiť
Capillary columns, HP-88
Dodávateľ: AGILENT HEWLETT PACKARD
112-88A7 112-8837LTM
HEWL112-88A7EA 2815.88 EUR
HEWL112-88A7 HEWL112-8837LTM
Capillary columns, HP-88
Kolony chromatografické
These columns are designed for separation of cis-trans fatty acid methyl esters (FAMES).
(88% -Cyanopropyl)aryl-polysiloxane, highly polar
Max. temperature: 250/320 °C