Tlačiť
Capillary columns, HP-PLOT Al₂O₃ S
Dodávateľ: AGILENT HEWLETT PACKARD
19095P-S23PT 19091P-S15PT 19091P-S12PT 19095P-S25PT 19091P-S12LTM
HEWL19095P-S23PTEA 1416.37 EUR
HEWL19095P-S23PT HEWL19091P-S15PT HEWL19091P-S12PT HEWL19095P-S25PT HEWL19091P-S12LTM
Capillary columns, HP-PLOT Al₂O₃ S
Kolony chromatografické
These columns are with middle range polarity. It is ideal for hydrocarbon analysis, and for resolving acetylene from butane and propylene from isobutane.
Aluminium oxide deactivated with sodium sulfate