Tlačiť

Anti-CD45.1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: A20]

Dodávateľ: Rockland Immunochemicals
ROCK200-526-N85EA 910.64 EUR
ROCK200-526-N85
Anti-CD45.1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: A20]
Protilátky
Anti-CD45.1 is useful for Flow Cytometry, Immunoprecipitation, and immunochemistry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze!

Type: Primary
Antigen: CD45.1
Clonality: Monoclonal
Clone: A20
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR