Tlačiť

Anti-CD19 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 6D5]

Dodávateľ: Rockland Immunochemicals
ROCK200-508-N81EA 910.64 EUR
ROCK200-508-N81
Anti-CD19 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 6D5]
Protilátky
Anti-CD19 is useful for Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze! Light sensitive!

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 6D5
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR