Tlačiť
Anti-SERF1A Rabbit Polyclonal Antibody (Biotin)
Dodávateľ: Novus Biologicals
NOVUNBP1-78393BEA 741.86 EUR
NOVUNBP1-78393B
Anti-SERF1A Rabbit Polyclonal Antibody (Biotin)
Protilátky
Type: Primary
Antigen: SERF1A (small EDRK-rich factor 1A (telomeric))
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human