Tlačiť

Anti-Hsp90B Rabbit Polyclonal Antibody

Dodávateľ: NOVUS BIOLOGICALS
NBP1-77561 NBP1-77561SS
NOVUNBP1-77561EA 632.08 EUR
NOVUNBP1-77561 NOVUNBP1-77561SS
Anti-Hsp90B Rabbit Polyclonal Antibody
Protilátky
Rabbit Polyclonal Hsp90B Antibody. Tested Applications: Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200

Type: Primary
Antigen: HSP90AB1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR