Tlačiť

Anti-KDM4A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodávateľ: Novus Biologicals
NOVUNBP1-49602HEA 801.83 EUR
NOVUNBP1-49602H
Anti-KDM4A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Type: Primary
Antigen: KDM4A (lysine (K)-specific demethylase 4A)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR