Tlačiť

Anti-CD44 Mouse Monoclonal Antibody [clone: 8E2F3]

Dodávateľ: NOVUS BIOLOGICALS
NBP1-47386 NBP1-47386SS
NOVUNBP1-47386EA 835.17 EUR
NOVUNBP1-47386 NOVUNBP1-47386SS
Anti-CD44 Mouse Monoclonal Antibody [clone: 8E2F3]
Protilátky
The CD44 Antibody (8E2F3) from Novus Biologicals is a mouse monoclonal antibody to CD44. This antibody reacts with human, mouse, rabbit (negative). The CD44 Antibody (8E2F3) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:200 - 1:1000

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 8E2F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rabbit (Negative)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR