Tlačiť

Anti-SULF2 Mouse Monoclonal Antibody [clone: 2B4]

Dodávateľ: NOVUS BIOLOGICALS
NBP1-36727 NBP1-36727SS
NOVUNBP1-36727EA 702.58 EUR
NOVUNBP1-36727 NOVUNBP1-36727SS
Anti-SULF2 Mouse Monoclonal Antibody [clone: 2B4]
Protilátky
The Sulfatase-2 / SULF2 Antibody (2B4) from Novus Biologicals is a mouse monoclonal antibody to Sulfatase-2 / SULF2. This antibody reacts with human, mouse. The Sulfatase-2 / SULF2 Antibody (2B4) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:250 - 1:500

Type: Primary
Antigen: SULF2
Clonality: Monoclonal
Clone: 2B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR