Tlačiť

Anti-AGT Rabbit Polyclonal Antibody

Dodávateľ: NOVUS BIOLOGICALS
NBP1-30027 NBP1-30027SS
NOVUNBP1-30027EA 742.64 EUR
NOVUNBP1-30027 NOVUNBP1-30027SS
Anti-AGT Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: AGT (angiotensinogen (serpin peptidase inhibitor, clade A, member 8))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR