Tlačiť

Anti-ACTB Rabbit Polyclonal Antibody (DyLight® 488)

Dodávateľ: Novus Biologicals
NOVUNB600-503GEA 888.01 EUR
NOVUNB600-503G
Anti-ACTB Rabbit Polyclonal Antibody (DyLight® 488)
Protilátky
Type: Primary
Antigen: ACTB (actin, beta)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR