Tlačiť

Anti-BIRC5 Rabbit Polyclonal Antibody (DyLight® 488)

Dodávateľ: Novus Biologicals
NOVUNB500-236GEA 787.13 EUR
NOVUNB500-236G
Anti-BIRC5 Rabbit Polyclonal Antibody (DyLight® 488)
Protilátky
Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR