Tlačiť

Anti-ROBO4 Rabbit Polyclonal Antibody

Dodávateľ: NOVUS BIOLOGICALS
NB110-58778 NB110-58778SS
NOVUNB110-58778EA 649.02 EUR
NOVUNB110-58778 NOVUNB110-58778SS
Anti-ROBO4 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: ROBO4 (roundabout, axon guidance receptor, homolog 4 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR