Tlačiť

Anti-Gankyrin Rabbit Polyclonal Antibody

Dodávateľ: NOVUS BIOLOGICALS
NB100-1834 NB100-1834SS
NOVUNB100-1834EA 632.08 EUR
NOVUNB100-1834 NOVUNB100-1834SS
Anti-Gankyrin Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Gankyrin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR