Tlačiť

Anti-IgG Donkey Polyclonal Antibody (CF™ 660C)

Dodávateľ: BIOTIUM
20051-1 20051
20051-1EA 171.44 EUR
20051-1 20051.
Anti-IgG Donkey Polyclonal Antibody (CF™ 660C)
Protilátky
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 660C Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR