Tlačiť
Anti-AKT1 (phospho S129) Rabbit Monoclonal Antibody [clone: EPR6150]
Dodávateľ: ABCAM
AB133458-100 AB133458-20UL AB133458-1ML
ABCAAB133458-100EA 543.12 EUR
ABCAAB133458-100 ABCAAB133458-20UL ABCAAB133458-1ML
Anti-AKT1 (phospho S129) Rabbit Monoclonal Antibody [clone: EPR6150]
Protilátky
Rabbit monoclonal [EPR6150] to AKT1 (phospho S129).
Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: AKT1
Clonality: Monoclonal
Clone: EPR6150
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human