Tlačiť

Anti-SNRPD3/Sm-D3 Rabbit Monoclonal Antibody [clone: EPR7676]

Dodávateľ: ABCAM
AB128857-100 AB128857-1ML
ABCAAB128857-100EA 714.8 EUR
ABCAAB128857-100 ABCAAB128857-1ML
Anti-SNRPD3/Sm-D3 Rabbit Monoclonal Antibody [clone: EPR7676]
Protilátky
Pozor:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: SNRPD3 (small nuclear ribonucleoprotein D3 polypeptide 18kDa)
Clonality: monoclonal
Clone: EPR7676
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR