Tlačiť

Anti-Glutathione S Transferase mu Rabbit Monoclonal Antibody [clone: EPR1936(2)]

Dodávateľ: ABCAM
AB108524-100 AB108524-1ML
ABCAAB108524-100EA 872.04 EUR
ABCAAB108524-100 ABCAAB108524-1ML
Anti-Glutathione S Transferase mu Rabbit Monoclonal Antibody [clone: EPR1936(2)]
Protilátky
Rabbit monoclonal [EPR1936(2)] to Glutathione S Transferase mu.

Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Glutathione S Transferase mu
Clonality: Monoclonal
Clone: EPR1936(2)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR