Tlačiť
Anti-EEF2/Elongation factor 2 Rabbit Monoclonal Antibody [clone: EP880Y]
Dodávateľ: ABCAM
AB75748-100 AB75748-1ML
ABCAAB75748-100EA 722.02 EUR
ABCAAB75748-100 ABCAAB75748-1ML
Anti-EEF2/Elongation factor 2 Rabbit Monoclonal Antibody [clone: EP880Y]
Protilátky
Rabbit monoclonal [EP880Y] to EEF2/Elongation factor 2.
Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: EEF2/Elongation factor 2
Clonality: Monoclonal
Clone: EP880Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat