Tlačiť

Anti-PKC delta Rabbit Monoclonal Antibody [clone: EPR1485Y]

Dodávateľ: ABCAM
AB53295-100 AB53295-1ML
ABCAAB53295-100EA 778.98 EUR
ABCAAB53295-100 ABCAAB53295-1ML
Anti-PKC delta Rabbit Monoclonal Antibody [clone: EPR1485Y]
Protilátky
Pozor:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.



Type: Primary
Antigen: PRKCD (protein kinase C, delta)
Clonality: monoclonal
Clone: EPR1485Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR