Tlačiť

Anti-KRT5 Rabbit Monoclonal Antibody [clone: EP1601Y]

Dodávateľ: ABCAM
AB52635-100 AB52635-1ML
ABCAAB52635-100EA 543.12 EUR
ABCAAB52635-100 ABCAAB52635-1ML
Anti-KRT5 Rabbit Monoclonal Antibody [clone: EP1601Y]
Protilátky
Rabbit monoclonal [EP1601Y] to Cytokeratin 5 - Cytoskeleton Marker.

Pozor: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: KRT5
Clonality: Monoclonal
Clone: EP1601Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR