Tlačiť

Anti-Histone H3 (phospho S10 + T11) Rabbit Monoclonal Antibody [clone: E173]

Dodávateľ: ABCAM
AB32107-100 AB32107-1ML AB32107-20UL
ABCAAB32107-100EA 850.38 EUR
ABCAAB32107-100 ABCAAB32107-1ML ABCAAB32107-20UL
Anti-Histone H3 (phospho S10 + T11) Rabbit Monoclonal Antibody [clone: E173]
Protilátky
Rabbit monoclonal [E173] to Histone H3 (phospho S10 + T11).

Pozor: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: E173
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR