Tlačiť

Anti-AIRE Rabbit Polyclonal Antibody

Dodávateľ: BIORBYT
ORB135062-100 ORB135062-200 ORB135062-50
BIRBORB135062-100EA 0 EUR
BIRBORB135062-100 BIRBORB135062-200 BIRBORB135062-50
Anti-AIRE Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: AIRE (autoimmune regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR