Tlačiť

Anti-MYC Rabbit Polyclonal Antibody

Dodávateľ: BIORBYT
ORB128408-100 ORB128408-200
BIRBORB128408-100EA 0 EUR
BIRBORB128408-100 BIRBORB128408-200
Anti-MYC Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: MYC (v-myc avian myelocytomatosis viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR