Tlačiť

Anti-KAT5 Rabbit Polyclonal Antibody

Dodávateľ: BIORBYT
ORB128244-100 ORB128244-200
BIRBORB128244-100EA 0 EUR
BIRBORB128244-100 BIRBORB128244-200
Anti-KAT5 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: KAT5 (K(lysine) acetyltransferase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR