Tlačiť

Anti-AIRE Rabbit Polyclonal Antibody

Dodávateľ: BIORBYT
ORB125403-100 ORB125403-200
BIRBORB125403-100EA 0 EUR
BIRBORB125403-100 BIRBORB125403-200
Anti-AIRE Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: AIRE (autoimmune regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR