Tlačiť

Anti-l-Myc Rabbit Polyclonal Antibody

Dodávateľ: BIORBYT
ORB101853-100 ORB101853-200
BIRBORB101853-100EA 0 EUR
BIRBORB101853-100 BIRBORB101853-200
Anti-l-Myc Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: l-Myc
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR