Tlačiť
Anti-ET-3 Rabbit Polyclonal Antibody
Dodávateľ: BIORBYT
ORB101459-100 ORB101459-200
BIRBORB101459-100EA 0 EUR
BIRBORB101459-100 BIRBORB101459-200
Anti-ET-3 Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: ET-3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human