Tlačiť

Anti-KLLN Rabbit Polyclonal Antibody

Dodávateľ: BIORBYT
ORB100624-100 ORB100624-200
BIRBORB100624-100EA 0 EUR
BIRBORB100624-100 BIRBORB100624-200
Anti-KLLN Rabbit Polyclonal Antibody
Protilátky
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: KLLN (killin, p53-regulated DNA replication inhibitor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR