Tlačiť
Anti-ET-1 Rabbit Polyclonal Antibody
Dodávateľ: BIORBYT
ORB13403-100 ORB13403-200
BIRBORB13403-100EA 0 EUR
BIRBORB13403-100 BIRBORB13403-200
Anti-ET-1 Rabbit Polyclonal Antibody
Protilátky
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: ET-1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat